Learn More
OHAUS™ Defender™ 3000 Stainless Steel
OHAUS™ Defender™ 3000 Stainless Steel is ideal for general weighing and simple counting applications when working in a damp environment. It features a simple, yet rugged stainless steel design, backlit LCD display, and built-in rechargeable battery operation.
Brand: OHAUS™ 83997637
This item is not returnable.
View return policy
Description
- The NEMA 4X/IP66 stainless steel indicator is built to endure tough industrial elements.
- The 58-hour rechargeable battery offers continuous operation for multi-day operations.
- Durable construction with a 304 stainless steel platform, stainless steel tubular frame, column and indicator bracket, and IP67 stainless steel load cell.
- Easy-to-read display and basic counting mode allow for simple operation for everyday basic industrial applications.
- Offered with NTEP or OIML/EC Type approvals (depending on region)
Specifications
58 Hours with Rechargeable Battery | |
IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, stainless steel column and wall bracket, 304 grade stainless steel platform with steel tubular frame, IP67 stainless steel load cell | |
Backlit LCD | |
D32XW60VR-EU | |
-10°C to +40°C | |
472 x 305 x 572 mm |
Easy access communication port with RS232 interface (included) | |
60 kg | |
10.000000 g | |
IP 65, NEMA 4X, IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, IP67 stainless steel load cell | |
Gram, Kilogram, Ounce, Pound, Pound:Ounce | |
14.500 kg |