missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helios Italquartz Deep and Shallow Crucibles, Transparent
Deep and shallow crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
65.70 € - 89.50 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
15858044
|
Helios Italquartz
63P00002 |
5 cc | 29 mm | 19 mm |
65.70 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
15868044
|
Helios Italquartz
63P00005 |
15 cc | 41 mm | 25 mm |
79.05 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
15878044
|
Helios Italquartz
63P00007 |
20 cc | 38 mm | 38 mm |
72.70 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
11784056
|
Helios Italquartz
63P00012 |
50 cc | 57 mm | 37 mm |
89.50 €
Each |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||
Description
- Transparent
- Deep and shallow forms available
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Transparent | |
Quartz | |
Crucible |