Learn More
DDX27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17087-1-AP-150UL
 View more versions of this product
Product Code. 16866924
  • 410.00 € / 150µL
Explore more special offers
Description
Description
La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU(OH) elements of newly RNA polymerase III.
Specifications
Specifications
DDX27
Polyclonal
Unconjugated
DDX27
DDX27, DEAD box protein 27, HSPC259, PP3241, RHLP, Rrp3p
Rabbit
Antigen Affinity Chromatography
RUO
228889, 362274, 55661
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q921N6, Q96GQ7
Ddx27
DDX27 Fusion Protein Ag9426
150 μL
Primary
Human, Mouse, Rat