Learn More
DAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22524-1-AP-20UL
 View more versions of this product
Product Code. 16851965
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson′s disease and drug abuse.
Specifications
Specifications
DAT
Polyclonal
Unconjugated
SLC6A3
DA transporter, DAT, DAT1, SLC6A3
Rabbit
Antigen Affinity Chromatography
RUO
13162, 24898, 6531
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23977, Q01959, Q61327
SLC6A3
DAT Fusion Protein Ag18297
20 μL
Primary
Mouse, Human, Rat