missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
252.00 € - 923.00 €
Chemical Identifiers
CAS | 87-69-4 |
---|---|
Molecular Formula | C4H6O6 |
Molecular Weight (g/mol) | 150.09 |
MDL Number | MFCD00064207 |
InChI Key | FEWJPZIEWOKRBE-UHFFFAOYNA-N |
Synonym | l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid |
PubChem CID | 444305 |
ChEBI | CHEBI:15671 |
SMILES | OC(C(O)C(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15527834
|
Avantor J.T.Baker
0386-01 |
500 g | Poly Bottle |
252.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15537834
|
Avantor J.T.Baker
0386-05 |
2.5 kg | Poly Bottle |
923.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
87-69-4 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
444305 | |
OC(C(O)C(O)=O)C(O)=O |
C4H6O6 | |
MFCD00064207 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
CHEBI:15671 |
Specifications
87-69-4 | |
MFCD00064207 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
(2R,3R)-2,3-dihydroxybutanedioic acid | |
444305 | |
150.09 | |
Crystalline | |
d-Tartaric Acid |
C4H6O6 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
OC(C(O)C(O)=O)C(O)=O | |
150.09 | |
CHEBI:15671 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 1.76kg |