missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Systec™ D-Series Horizontal Benchtop Autoclaves, Systec DX
Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ D-Series Horizontal Benchtop Autoclaves feature a compact and space-saving design. For all laboratory applications including sophisticated sterilization processes.
Specifications
Design | Horizontal benchtop |
---|---|
For Use With (Application) | For all laboratory applications even for sophisticated state-of-the-art sterilization processes |
Product Type | D-Series Benchtop Autoclave |
Display | LCD display and fully automatic microprocessor control |
Description
Standard Features- Integrated, separate steam generator
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- Temperature and pressure range 140°C, 4 bar (except for Systec D-23)
- LCD display and fully automatic microprocessor control
- Number of sterilization programs: Up to 25
- Code-secured access rights for changing parameters and further safety-relevant intervention
- Internal memory for storing up to 500 sterilization cycles
- Timer for starting programs
- Autofill: automatic demineralized water feed for steam generation
- Flexible PT-100 temperature sensor
- Additional temperature sensor in condense exhaust
- Temperature holding function for liquids after program finish
- Special program for Durham tubes
- Calculation of F0 value
- Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
- Water-cooled steam exhaust, thermostatically controlled
- Programmable automatic door-opening on completion of program
- RS-232 and RS-485 interfaces for external data transmission (network-compatible)
Available options
- Extension of temperature and pressure ranges to 150°C / 5 bar (from chamber volume 65 liters and more)
Options for process optimization
- Rapid cooling for efficient and safe cooling of liquids
- Vacuum system for validatable sterilization of solids and waste materials in disposal bags
- Superdry: for drying solids (only in combination with optional vacuum system)
- Exhaust filtration (including condensate inactivation) for safe sterilization of hazardous biological substances
Options for documentation
- Integrated printer for batch documentation
- PC software for comprehensive documentation
- Documentation SD: data storage on SD card for up to 10,000 sterilization cycles and transmission of data to a PC
Specifications
Horizontal benchtop | |
For all laboratory applications even for sophisticated state-of-the-art sterilization processes | |
D-Series Benchtop Autoclave | |
LCD display and fully automatic microprocessor control |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title