Learn More
D-(-)-Quinic acid, 98%, Thermo Scientific Chemicals
21.00 € - 48.30 €
Chemical Identifiers
CAS | 77-95-2 |
---|---|
Molecular Formula | C7H12O6 |
Molecular Weight (g/mol) | 192.17 |
MDL Number | MFCD00003864 |
InChI Key | AAWZDTNXLSGCEK-UHFFFAOYNA-N |
Synonym | quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi |
PubChem CID | 6508 |
IUPAC Name | (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid |
SMILES | OC1CC(O)(CC(O)C1O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398216
|
Thermo Scientific Alfa Aesar
L15238.06 |
5 g |
21.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308226
|
Thermo Scientific Alfa Aesar
L15238.14 |
25 g |
48.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.
Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
77-95-2 | |
192.17 | |
AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
6508 | |
OC1CC(O)(CC(O)C1O)C(O)=O |
C7H12O6 | |
MFCD00003864 | |
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid |
Specifications
77-95-2 | |
Odorless | |
MFCD00003864 | |
14,8059 | |
Soluble in water (400g/L),methanol and dimethyl sulfoxide. | |
−44° (c=20 in Water) | |
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid | |
6508 | |
98% |
162°C to 167°C | |
C7H12O6 | |
2212412 | |
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
OC1CC(O)(CC(O)C1O)C(O)=O | |
192.17 | |
192.17 | |
D-(-)-Quinic acid |
Safety and Handling
GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 201-072-8
missing translation for 'rtecsNumber' : GU8650000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only