missing translation for 'onlineSavingsMsg'
Learn More

D-Psicose, 98%, Thermo Scientific Chemicals

322.61 € - 1155.75 €

Chemical Identifiers

CAS 551-68-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00083478
InChI Key BJHIKXHVCXFQLS-PUFIMZNGSA-N
Synonym erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
PubChem CID 90008
ChEBI CHEBI:27605
IUPAC Name (3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15355068
View Documents
Thermo Scientific Acros
456270010
1 g
322.61 €
1g
Please to purchase this item. Need a web account? Register with us today!
15365068
View Documents
Thermo Scientific Acros
456270050
5 g
1155.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

551-68-8
180.16
BJHIKXHVCXFQLS-PUFIMZNGSA-N
90008
(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
MFCD00083478
erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
CHEBI:27605
C(C(C(C(C(=O)CO)O)O)O)O

Specifications

551-68-8
White
Glass Bottle
MFCD00083478
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in acetone
C(C(C(C(C(=O)CO)O)O)O)O
180.16
CHEBI:27605
98%
D-Psicose
109.0°C
97.5% min. (ELSD) (HPLC)
C6H12O6
erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
BJHIKXHVCXFQLS-PUFIMZNGSA-N
(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
90008
180.16
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-999-7

Product Identifier
  • D-Psicose

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.