missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Pinitol, 95%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
Solubility
Soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
Solubility
Soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Specifications
Melting Point | 178°C to 185°C |
Quantity | 5 g |
Merck Index | 14,10361 |
Solubility Information | Soluble in water. |
Optical Rotation | +60° (c=1 in Water) |
Formula Weight | 194.18 |
Percent Purity | 95% |
Chemical Name or Material | D-Pinitol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'