Learn More
Thermo Scientific Chemicals D-(+)-Maltose monohydrate, 95%
42.31 € - 121.61 €
Chemical Identifiers
CAS | 6363-53-7 |
---|---|
Molecular Formula | C12H24O12 |
Molecular Weight (g/mol) | 360.31 |
MDL Number | MFCD00149343 |
InChI Key | HBDJFVFTHLOSDW-UHFFFAOYNA-N |
Synonym | d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra |
IUPAC Name | 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate |
SMILES | O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467727
|
Thermo Scientific Chemicals
A16266.22 |
100 g |
42.31 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477727
|
Thermo Scientific Chemicals
A16266.36 |
500 g |
121.61 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-(+)-Maltose monohydrate, is commonly found in foods and commonly utilized in brewing processes. It is also used in various culture media in the cell and tissue culture applications. D-(+)-Maltose Monohydrate is used as a substrate for α-glucosidase. It is also used as a substrate for the identification, differentiation and characterization of enzymes such as maltase(s); maltose α-D-glucosyltransferase(s); maltose-transporting ATPase(s); maltose O-acetyltransferase(s) and maltose epimerase(s) and phosphorylase(s). D-Maltose is used to study maltose-binding proteins and disaccharide transport systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(+)-Maltose monohydrate, is commonly found in foods and commonly utilized in brewing processes. It is also used in various culture media in the cell and tissue culture applications. D-(+)-Maltose Monohydrate is used as a substrate for α-glucosidase. It is also used as a substrate for the identification, differentiation and characterization of enzymes such as maltase(s); maltose α-D-glucosyltransferase(s); maltose-transporting ATPase(s); maltose O-acetyltransferase(s) and maltose epimerase(s) and phosphorylase(s). D-Maltose is used to study maltose-binding proteins and disaccharide transport systems.
Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in ether
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
Chemical Identifiers
6363-53-7 | |
360.31 | |
HBDJFVFTHLOSDW-UHFFFAOYNA-N | |
2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate |
C12H24O12 | |
MFCD00149343 | |
d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra | |
O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O |
Specifications
6363-53-7 | |
95% | |
MFCD00149343 | |
14,5714 | |
Soluble in water; Slightly soluble in alcohol; Insoluble in ether | |
O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O | |
360.31 | |
95% |
∽120°C (decomposition) | |
C12H24O12 | |
5784659 | |
d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra | |
HBDJFVFTHLOSDW-UHFFFAOYNA-N | |
2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate | |
360.32 (342.30 Anhydrous) | |
D-(+)-Maltose monohydrate |
Safety and Handling
missing translation for 'einecsNumber' : 200-716-5
missing translation for 'rtecsNumber' : OO5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only