missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic acid 1-tert-butyl ester, 98%, Thermo Scientific Chemicals

28.00 € - 358.00 €

Chemical Identifiers

CAS 25456-76-2
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD09842819
InChI Key QVAQMUAKTNUNLN-ZCFIWIBFSA-N
Synonym h-d-glu-otbu, r-4-amino-5-tert-butoxy-5-oxopentanoic acid, 4r-4-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamic acid 1-tert-butyl ester, r-4-amino-5-tert-butoxy-5-oxopentanoicacid
PubChem CID 7019726
IUPAC Name (4R)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(=O)O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15449958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63318.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15429958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63318.03
1 g
84.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
15439958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63318.06
5 g
358.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

25456-76-2
203.238
QVAQMUAKTNUNLN-ZCFIWIBFSA-N
7019726
CC(C)(C)OC(=O)C(CCC(=O)O)N
C9H17NO4
MFCD09842819
h-d-glu-otbu, r-4-amino-5-tert-butoxy-5-oxopentanoic acid, 4r-4-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamic acid 1-tert-butyl ester, r-4-amino-5-tert-butoxy-5-oxopentanoicacid
(4R)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid

Specifications

25456-76-2
C9H17NO4
h-d-glu-otbu, r-4-amino-5-tert-butoxy-5-oxopentanoic acid, 4r-4-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamic acid 1-tert-butyl ester, r-4-amino-5-tert-butoxy-5-oxopentanoicacid
QVAQMUAKTNUNLN-ZCFIWIBFSA-N
(4R)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
7019726
98%
D-Glutamic acid 1-tert-butyl ester
White
MFCD09842819
Slightly soluble in water.
CC(C)(C)OC(=O)C(CCC(=O)O)N
203.238
203.24
Powder
Vidéos
Sécurité et traitement

Sécurité et traitement

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

FDS
Documents

Documents

Product Certifications

RUO – Research Use Only