missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Galactose, 99+%, Thermo Scientific Chemicals

Product Code. 10649283
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg

D-Galactose, CAS # 59-23-4, is a right-handed conformation of a monosaccharide that is grouped as an aldohexose, and the molecule may be useful in study of aging, inflammation, and metabolic disorders.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Galactose is a monosaccharide and aldohexose found in most organisms
  • Galactose is an epimer of glucose and can be converted to glucose through the Leloir pathway
  • It also plays a role as a metabolite in mice and E. coli
  • Galactose is less stable than glucose and rapid conversion often occurs
  • Galactose may also be incorporated into glycoconjugates

Applications

  • Metabolism of galactose may be relevant to the study of aging, inflammation, and metabolic disorders
  • It is intended for a variety of applications from in vitro metabolic assays to HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00063833

Specifications

Melting Point 165.0°C to 168.0°C
Color White
Density 6.2
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 5 kg
Beilstein 31, 295
Merck Index 15, 4365
Specific Rotation Condition + 79.75 (20.00°C c=10,H2O,NH3)
Specific Rotation + 79.75
Formula Weight 180.16
Vapor Pressure Negligible
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D(+)-Galactose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.