Learn More
Thermo Scientific Chemicals D-Fructose, 99%
30.95 € - 234.17 €
Chemical Identifiers
CAS | 57-48-7 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
MDL Number | MFCD00148910 |
InChI Key | BJHIKXHVCXFQLS-UYFOZJQFSA-N |
Synonym | d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 |
PubChem CID | 5984 |
ChEBI | CHEBI:48095 |
IUPAC Name | (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
SMILES | C(C(C(C(C(=O)CO)O)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453308
|
Thermo Scientific Chemicals
A17718.30 |
250 g |
30.95 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443308
|
Thermo Scientific Chemicals
A17718.0B |
1000 g |
67.54 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463308
|
Thermo Scientific Chemicals
A17718.0I |
5000 g |
234.17 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.
Solubility
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
57-48-7 | |
180.156 | |
BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
5984 | |
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
C6H12O6 | |
MFCD00148910 | |
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 | |
CHEBI:48095 | |
C(C(C(C(C(=O)CO)O)O)O)O |
Specifications
57-48-7 | |
1.59 | |
C6H12O6 | |
1239004 | |
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 | |
BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
5984 | |
180.16 | |
D-Fructose |
∽105°C to 110°C | |
99% | |
MFCD00148910 | |
14,4273 | |
Soluble in water. | |
C(C(C(C(C(=O)CO)O)O)O)O | |
180.156 | |
CHEBI:48095 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 200-333-3
missing translation for 'rtecsNumber' : LS7120000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.