missing translation for 'onlineSavingsMsg'
Learn More

D-Epoxone, 98%, Thermo Scientific Chemicals

Product Code. 11399565
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18422-53-2
Molecular Formula C12H18O6
Molecular Weight (g/mol) 258.27
MDL Number MFCD00063383
InChI Key IVWWFWFVSWOTLP-RDNCKJRISA-N
Synonym 3ar,4'r,7ar-2,2,2',2'-tetramethyldihydrospiro 1,3 dioxolo 4,5-c pyran-6,4'-1,3 dioxolan-7 7ah-one
PubChem CID 118990039
IUPAC Name (4S,7'aR)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,6'-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran]-7'-one
SMILES CC1(OCC2(O1)C(=O)C3C(CO2)OC(O3)(C)C)C

Specifications

Melting Point 101°C to 103°C
Quantity 5 g
Beilstein 1319073
Solubility Information Soluble in chloroform and methanol.
Formula Weight 258.27
Percent Purity 98%
Chemical Name or Material D-Epoxone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'