missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-δ-Tocopherol, 93%, Thermo Scientific Chemicals
197.99 € - 753.75 €
Chemical Identifiers
CAS | 119-13-1 |
---|---|
Molecular Formula | C27H46O2 |
Molecular Weight (g/mol) | 402.66 |
MDL Number | MFCD20486794 |
InChI Key | GZIFEOYASATJEH-VHFRWLAGSA-N |
Synonym | delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r |
PubChem CID | 12444418 |
SMILES | CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10391641
|
Thermo Scientific Acros
421051000 |
100 mg | Glass bottle |
197.99 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936881
|
Thermo Scientific Acros
421055000 |
500 mg | Glass bottle |
753.75 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
119-13-1 | |
402.66 | |
GZIFEOYASATJEH-VHFRWLAGSA-N | |
12444418 |
C27H46O2 | |
MFCD20486794 | |
delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1 |
Specifications
119-13-1 | |
>93°C | |
91% min. (GC) | |
MFCD20486794 | |
delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r | |
GZIFEOYASATJEH-VHFRWLAGSA-N | |
(2S)-2,8-dimethyl-2-[(4S,8S)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol | |
12444418 | |
93% | |
D-δ-Tocopherol |
Brown to Yellow | |
Authentic | |
C27H46O2 | |
15,9656 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone, chloroform, ether,, oils, fats | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1 | |
402.66 | |
402.65 | |
Viscous Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 204-299-
Product Identifier
- D-delta-Tocopherol
RUO – Research Use Only