missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2D6 Mouse anti-Human, Clone: 8B11D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60297-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant. The gene is highly polymorphic in the population; certain alleles result in the poor metabolizer phenotype, characterized by a decreased ability to metabolize the enzyme′s substrates. The gene is located near two cytochrome P450 pseudogenes on chromosome 22q13.1. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
CYP2D6 | |
Monoclonal | |
1.85 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10635 | |
CYP2D6 | |
CYP2D6 Fusion Protein Ag12605 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
8B11D12 | |
Unconjugated | |
CYP2D6 | |
CPD6, CYP2D, CYP2D6, CYP2DL1, Cytochrome P450 2D6, P450 DB1, P450C2D, P450DB1 | |
Mouse | |
Protein A | |
RUO | |
1565 | |
-20°C | |
Liquid |