missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyp2c7 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 410.00 €
Specifications
Antigen | Cyp2c7 |
---|---|
Concentration | 380 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816925
|
Proteintech
55452-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806925
|
Proteintech
55452-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cyp2c7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of rat Cyp2c7. This antibody recognizes mouse, rat antigen. The Cyp2c7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Cyp2c7 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Mouse, Rat | |
IgG |
380 μg/mL | |
Polyclonal | |
Rabbit | |
29298 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title