Learn More
Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
96.00 € - 292.00 €
Chemical Identifiers
CAS | 23719-80-4 |
---|---|
Molecular Formula | C3H5BrMg |
Molecular Weight (g/mol) | 145.28 |
MDL Number | MFCD00672094 |
InChI Key | QUPLLJGAJVXXBW-UHFFFAOYSA-M |
Synonym | cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent |
PubChem CID | 11116189 |
IUPAC Name | magnesium;cyclopropane;bromide |
SMILES | Br[Mg]C1CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366759
|
Thermo Scientific Alfa Aesar
H26273.AC |
25 mL | |||||||
11376759
|
Thermo Scientific Alfa Aesar
H26273.AE |
100 mL |
292.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.
Solubility
Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.
Chemical Identifiers
23719-80-4 | |
145.28 | |
QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
11116189 | |
Br[Mg]C1CC1 |
C3H5BrMg | |
MFCD00672094 | |
cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent | |
magnesium;cyclopropane;bromide |
Specifications
23719-80-4 | |
−17°C (1°F) | |
MFCD00672094 | |
Air and moisture sensitive; May form a precipitate | |
Soluble in water (17mg/ml),(25°C). Soluble in organic solvents. | |
Br[Mg]C1CC1 | |
145.28 | |
145.28 | |
Cyclopropylmagnesium bromide |
0.968 | |
C3H5BrMg | |
UN2924 | |
cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent | |
QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
magnesium;cyclopropane;bromide | |
11116189 | |
0.5 to 0.7M in THF |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335-H336-H351
missing translation for 'dotInformation' : Hazard Class: 4.3; Packaging Group: I
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only