missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylamine, 99%, Thermo Scientific Chemicals
35.38 € - 500.49 €
Chemical Identifiers
CAS | 765-30-0 |
---|---|
Molecular Formula | C3H7N |
Molecular Weight (g/mol) | 57.09 |
InChI Key | HTJDQJBWANPRPF-UHFFFAOYSA-N |
Synonym | cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine |
PubChem CID | 69828 |
ChEBI | CHEBI:34660 |
IUPAC Name | cyclopropanamine |
SMILES | C1CC1N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163250
|
Thermo Scientific Acros
154880100 |
10 mL | Glass bottle |
35.38 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205630
|
Thermo Scientific Acros
154880500 |
50 mL | Glass bottle |
129.65 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995191
|
Thermo Scientific Acros
154882500 |
250 mL | Glass bottle |
500.49 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
765-30-0 | |
57.09 | |
cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine | |
CHEBI:34660 | |
C1CC1N |
C3H7N | |
HTJDQJBWANPRPF-UHFFFAOYSA-N | |
69828 | |
cyclopropanamine |
Specifications
765-30-0 | |
Colorless | |
49.0°C to 50.0°C | |
Authentic | |
C3H7N | |
1.4190 to 1.4220 | |
0.824 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
C1CC1N | |
57.09 | |
CHEBI:34660 | |
99% | |
Cyclopropylamine |
-50.0°C | |
0.8240g/mL | |
−25°C | |
99% min. (GC) | |
C3H5NH2 | |
12,3 | |
cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine | |
HTJDQJBWANPRPF-UHFFFAOYSA-N | |
cyclopropanamine | |
69828 | |
57.09 | |
Volatile Liquid |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-142-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cyclopropylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title