missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Product Code. 15400368
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

Solubility
Sparingly Soluble (0.069 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81166-84-9
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.29
MDL Number MFCD00671355
InChI Key IYQSVLJDYLPMCX-UHFFFAOYSA-N
Synonym cyclopropyl trimethylsilyl acetylene, cyclopropylethynyl trimethylsilane, 2-cyclopropylethynyl trimethylsilane, cyclopropylethynyl trimethyl silane, silane, cyclopropylethynyl trimethyl, cyclopropane, 2-trimethylsilyl ethynyl, silane, cyclopropylethynyl trimethyl-9ci, timtec-bb sbb009013, trimethylsilylethynylcyclopropane, cyclopropylethynyl-trimethylsilane
PubChem CID 2758018
SMILES C[Si](C)(C)C#CC1CC1

Specifications

Boiling Point 83°C (80mmHg)
Quantity 25 g
UN Number UN1993
Flash Point 25°C (77°F)
Solubility Information Sparingly Soluble (0.069g/L) (25°C).
IUPAC Name (2-cyclopropylethynyl)trimethylsilane
Formula Weight 138.28
Percent Purity 97%
Refractive Index 1.454
Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'