Learn More
Cyclopentanone, 99%, Thermo Scientific Chemicals
30.10 € - 161.00 €
Chemical Identifiers
CAS | 120-92-3 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 84.12 |
MDL Number | MFCD00001409 |
InChI Key | BGTOWKSIORTVQH-UHFFFAOYSA-N |
Synonym | adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy |
PubChem CID | 8452 |
ChEBI | CHEBI:16486 |
IUPAC Name | cyclopentanone |
SMILES | O=C1CCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374529
|
Thermo Scientific Alfa Aesar
A14222.AK |
250 mL |
30.10 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432327
|
Thermo Scientific Alfa Aesar
A14222.AP |
500 mL |
50.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442327
|
Thermo Scientific Alfa Aesar
A14222.0F |
2500 mL |
161.00 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclopentanone is used as an intermediate in the synthesis of rubber adhesives, synthetic resins, pharmaceuticals and biologically active compounds. It acts as precursor for the preparation of cyclopentamine and also pentethylcyclanone, cyclopentobarbital. It is a useful laboratory reagent and is used as thinner for epoxies. It can also be used as a solvent in paint and varnish removers and for electronic applications. As a dry cleaning agent, it is used for oil extraction. It is also involved in the preparation of cyclopentanone derivatives like cyclopenylamine and cyclopentanol which find application in the perfume industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopentanone is used as an intermediate in the synthesis of rubber adhesives, synthetic resins, pharmaceuticals and biologically active compounds. It acts as precursor for the preparation of cyclopentamine and also pentethylcyclanone, cyclopentobarbital. It is a useful laboratory reagent and is used as thinner for epoxies. It can also be used as a solvent in paint and varnish removers and for electronic applications. As a dry cleaning agent, it is used for oil extraction. It is also involved in the preparation of cyclopentanone derivatives like cyclopenylamine and cyclopentanol which find application in the perfume industry.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong reducing agents, strong oxidizing agents and strong bases.
Chemical Identifiers
120-92-3 | |
84.12 | |
BGTOWKSIORTVQH-UHFFFAOYSA-N | |
8452 | |
cyclopentanone |
C5H8O | |
MFCD00001409 | |
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy | |
CHEBI:16486 | |
O=C1CCCC1 |
Specifications
120-92-3 | |
0.949 | |
30°C (86°F) | |
C5H8O | |
MFCD00001409 | |
605573 | |
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy | |
BGTOWKSIORTVQH-UHFFFAOYSA-N | |
cyclopentanone | |
8452 | |
84.12 | |
Cyclopentanone |
-51°C | |
130°C to 131°C | |
Petrol-like | |
1.437 | |
UN2245 | |
14,2743 | |
Slightly miscible with water. | |
O=C1CCCC1 | |
84.12 | |
CHEBI:16486 | |
99% |
Safety and Handling
GHS H Statement
H226-H315-H319
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANONE
missing translation for 'einecsNumber' : 204-435-9
missing translation for 'rtecsNumber' : GY4725000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only