missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, Reagent Grade, 98%, Honeywell

Product Code. 15625610
Click to view available options
Quantity:
100 mL
500 mL
2 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Melting Point -94°C
Color Colorless
Density 0.751g/mL at 25°C
Boiling Point 50°C
Flash Point −20°C (−4°F)
Ignition Point 682°F
Odor Solvent like
Packaging Glass Bottle
Quantity 500 mL
UN Number UN1146
Beilstein 1900195
Formula Weight 70.13g/mol
Vapor Pressure 5.3psi (21°C), 9.9psi (38°C)
Percent Purity 98%
Chemical Name or Material Cyclopentane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.