missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, 98%, Thermo Scientific Chemicals

Product Code. 11479946
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Cyclooctanone is a aliphatic cycle that exhibited inhibitory activity towards aldosterone synthase, a promising therapeutic target for the treatment of cardiovascular diseases related to abnormally high aldosterone level. It is also used in the synthesis of 14-membered lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 20°C 15g/L. Soluble in acetone, alcohol, chloroform, methanol and benzene.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-49-8
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00001754
InChI Key IIRFCWANHMSDCG-UHFFFAOYSA-N
Synonym cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
PubChem CID 10403
IUPAC Name cyclooctanone
SMILES C1CCCC(=O)CCC1

Specifications

Melting Point 37°C to 44°C
Density 0.96
Boiling Point 195°C to 197°C
Flash Point 72°C (161°F)
Odor Pleasant
Quantity 100 g
UN Number UN1759
Beilstein 1280738
Solubility Information Soluble in water at 20°C 15g/L. Soluble in acetone,alcohol,chloroform,methanol and benzene.
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material Cyclooctanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'