Learn More
Cyclohexanemethylamine, 98%, Thermo Scientific Chemicals
38.59 € - 115.58 €
Chemical Identifiers
CAS | 3218-02-8 |
---|---|
Molecular Formula | C7H15N |
Molecular Weight (g/mol) | 113.20 |
MDL Number | MFCD00001520 |
InChI Key | AVKNGPAMCBSNSO-UHFFFAOYSA-N |
Synonym | cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049 |
PubChem CID | 76688 |
IUPAC Name | cyclohexylmethanamine |
SMILES | C1CCC(CC1)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312546
|
Thermo Scientific Alfa Aesar
L06501.06 |
5 g |
38.59 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322546
|
Thermo Scientific Alfa Aesar
L06501.14 |
25 g |
115.58 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclohexanemethylamine was used in the synthesis of Schiff?s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexanemethylamine was used in the synthesis of Schiff′s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
3218-02-8 | |
113.20 | |
AVKNGPAMCBSNSO-UHFFFAOYSA-N | |
76688 | |
C1CCC(CC1)CN |
C7H15N | |
MFCD00001520 | |
cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049 | |
cyclohexylmethanamine |
Specifications
3218-02-8 | |
0.871 | |
43°C (109°F) | |
1.462 | |
UN2734 | |
Air Sensitive | |
Slightly soluble in water. | |
C1CCC(CC1)CN | |
113.20 | |
113.2 | |
Cyclohexanemethylamine |
-8°C | |
161°C to 163°C | |
C7H15N | |
MFCD00001520 | |
635751 | |
cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049 | |
AVKNGPAMCBSNSO-UHFFFAOYSA-N | |
cyclohexylmethanamine | |
76688 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 221-741-8
missing translation for 'rtecsNumber' : GV6128000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.