missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™

Product Code. 15604230
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Density 0.779g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.09
Flash Point −18°C (−.4°F)
Ignition Point 500°F
Vapor Pressure 168.8 mmHg (37.7°C), 77 mmHg (20°C)
UN Number UN1145
Formula Weight 84.16
Percent Purity ≥99.5% (GC)
Chemical Name or Material Cyclohexane
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cyclohexane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.