missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications

Specifications
Specifications
Melting Point | 6.4°C |
Density | 0.779g/mL at 25°C |
Boiling Point | 81°C |
Quantity | 20 L |
Explosion Limit | 0.09 |
Flash Point | −18°C (−.4°F) |
Ignition Point | 500°F |
Packaging | Stainless Steel Can |
Physical Form | Liquid |
Refractive Index | n20/D 1.426(lit.) |
Show More |
Safety and Handling




Product Identifier
- Cyclohexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cyclohexane, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction