Learn More
Cycloheptylamine, 99%, Thermo Scientific Chemicals
39.30 € - 114.20 €
Chemical Identifiers
CAS | 5452-35-7 |
---|---|
Molecular Formula | C7H15N |
Molecular Weight (g/mol) | 113.2 |
MDL Number | MFCD00004153 |
InChI Key | VXVVUHQULXCUPF-UHFFFAOYSA-N |
Synonym | cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, acmc-209lh8 |
PubChem CID | 2899 |
IUPAC Name | cycloheptanamine |
SMILES | C1CCCC(CC1)N |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5452-35-7 | |
113.2 | |
VXVVUHQULXCUPF-UHFFFAOYSA-N | |
2899 | |
C1CCCC(CC1)N |
C7H15N | |
MFCD00004153 | |
cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, acmc-209lh8 | |
cycloheptanamine |
Specifications
5452-35-7 | |
0.8900g/mL | |
42°C | |
98.5% min. (GC) | |
C7H13NH2 | |
MFCD00004153 | |
0.89 | |
Solubility in water: soluble. | |
C1CCCC(CC1)N | |
113.2 | |
113.2 | |
Liquid |
Colorless to Yellow | |
54.0°C (11.0 mmHg) | |
Authentic | |
C7H15N | |
1.4710 to 1.4730 | |
12, 8 | |
cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, acmc-209lh8 | |
VXVVUHQULXCUPF-UHFFFAOYSA-N | |
cycloheptanamine | |
2899 | |
99% | |
Cycloheptylamine, 99% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 226-693-1
missing translation for 'tsca' : TSCA
- Cycloheptylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.