Learn More
Cycloheptene, 95%, Thermo Scientific Chemicals
103.00 € - 341.00 €
Chemical Identifiers
CAS | 628-92-2 |
---|---|
Molecular Formula | C7H12 |
Molecular Weight (g/mol) | 96.17 |
MDL Number | MFCD00004156 |
InChI Key | ZXIJMRYMVAMXQP-UHFFFAOYSA-N |
Synonym | cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure |
PubChem CID | 12363 |
IUPAC Name | cycloheptene |
SMILES | C1CCC=CCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10155850
|
Thermo Scientific Acros
304260050 |
5 g |
103.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10105330
|
Thermo Scientific Acros
304260250 |
25 g |
341.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
628-92-2 | |
96.17 | |
ZXIJMRYMVAMXQP-UHFFFAOYSA-N | |
12363 | |
C1CCC=CCC1 |
C7H12 | |
MFCD00004156 | |
cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure | |
cycloheptene |
Specifications
628-92-2 | |
Colorless | |
114.6°C | |
Authentic | |
Glass bottle | |
C7H12 | |
MFCD00004156 | |
0.82 | |
Solubility in water: immiscible with water. Other solubilities: soluble in ether,ethanol,benzene | |
C1CCC=CCC1 | |
96.17 | |
96.17 | |
Liquid |
-56°C | |
0.8200g/mL | |
-006°C | |
94% min. (GC) | |
C7H12 | |
1.4570 to 1.459 | |
05, 65 | |
cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure | |
ZXIJMRYMVAMXQP-UHFFFAOYSA-N | |
cycloheptene | |
12363 | |
95% | |
Cycloheptene, 90% |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-060-4
RUO – Research Use Only