Learn More
Cycloheptanemethylamine, 98+%, Thermo Scientific Chemicals
57.10 € - 202.00 €
Chemical Identifiers
CAS | 4448-77-5 |
---|---|
Molecular Formula | C8H17N |
Molecular Weight (g/mol) | 127.231 |
MDL Number | MFCD00021674 |
InChI Key | CAOQEOHEZKVYOJ-UHFFFAOYSA-N |
Synonym | cycloheptanemethylamine, cycloheptylmethylamine, cycloheptanemethanamine, acmc-20agqt, cycloheptylmethyl amine, cycloheptane methylamine, aminomethyl cycloheptane, n-cycloheptylmethyl-amine, aminomethylcycloheptane |
PubChem CID | 78194 |
IUPAC Name | cycloheptylmethanamine |
SMILES | C1CCCC(CC1)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353636
|
Thermo Scientific Alfa Aesar
L14659.03 |
1 g |
57.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363636
|
Thermo Scientific Alfa Aesar
L14659.06 |
5 g |
202.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible or difficult to mix with water.
Notes
Air Sensitive, Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
4448-77-5 | |
127.231 | |
CAOQEOHEZKVYOJ-UHFFFAOYSA-N | |
78194 | |
C1CCCC(CC1)CN |
C8H17N | |
MFCD00021674 | |
cycloheptanemethylamine, cycloheptylmethylamine, cycloheptanemethanamine, acmc-20agqt, cycloheptylmethyl amine, cycloheptane methylamine, aminomethyl cycloheptane, n-cycloheptylmethyl-amine, aminomethylcycloheptane | |
cycloheptylmethanamine |
Specifications
4448-77-5 | |
98°C to 101°C (35 mmHg) | |
C8H17N | |
MFCD00021674 | |
2958734 | |
cycloheptanemethylamine, cycloheptylmethylamine, cycloheptanemethanamine, acmc-20agqt, cycloheptylmethyl amine, cycloheptane methylamine, aminomethyl cycloheptane, n-cycloheptylmethyl-amine, aminomethylcycloheptane | |
CAOQEOHEZKVYOJ-UHFFFAOYSA-N | |
cycloheptylmethanamine | |
78194 | |
≥98% |
0.884 | |
70°C (158°F) | |
1.474 | |
UN2735 | |
Air Sensitive | |
Not miscible or difficult to mix with water. | |
C1CCCC(CC1)CN | |
127.231 | |
127.23 | |
Cycloheptanemethylamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 224-692-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only