missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclododecanone, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 59.0°C to 61.0°C |
Color | Colorless to White |
Boiling Point | 277.0°C |
Flash Point | 118°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Linear Formula | C12H22(=O) |
Quantity | 500 g |
Beilstein | 07, II, 48 |
Show More |
Safety and Handling
Product Identifier
- Cyclododecanone
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'