missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanone, 98+%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 1191-95-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00001332
InChI Key SHQSVMDWKBRBGB-UHFFFAOYSA-N
Synonym unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
PubChem CID 14496
IUPAC Name cyclobutanone
SMILES O=C1CCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10560191
View Documents
Thermo Scientific Acros
1 g Glass bottle N/A
Please to purchase this item. Need a web account? Register with us today!
10030890
View Documents
Thermo Scientific Acros
5 g Glass bottle N/A
Please to purchase this item. Need a web account? Register with us today!
11964831
View Documents
Thermo Scientific Acros
25 g Glass bottle N/A
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1191-95-3
70.09
SHQSVMDWKBRBGB-UHFFFAOYSA-N
14496
O=C1CCC1
C4H6O
MFCD00001332
unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
cyclobutanone

Specifications

1191-95-3
Colorless to Yellow
99.0°C
Authentic
C4H6O
1.4205 to 1.4225
07,5
unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
SHQSVMDWKBRBGB-UHFFFAOYSA-N
cyclobutanone
14496
98+%
Cyclobutanone
-50.9°C
0.9300g/mL
10°C
98% min. (GC)
C4H6(=O)
MFCD00001332
0.93
Solubility in water: insoluble
O=C1CCC1
70.09
70.09
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 214-745-6

Product Identifier
  • Cyclobutanone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.