missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55004-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to CCNB1. The observed 55-60 kDa band in western blotting maybe due to phosphorylation.
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predomitly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.Specifications
Cyclin B1 | |
Polyclonal | |
Unconjugated | |
CCNB1 | |
CCNB, CCNB1, cyclin B1, G2/mitotic specific cyclin B1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
268697, 891 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14635, P24860 | |
CCNB1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |