missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Cy5.5 Hydrazide
Brand: Cytiva PA15620
This item is not returnable.
View return policy
Description
Specifications
100% Cy5.5 (hydrazide) | |
5 mg | |
Foil packs, each containing 1, 5 or 25mg dried dye | |
Soluble in cold water and hot water | |
Cy5.5 |
For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications | |
Cy5.5 Hydrazide | |
1128.42 | |
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. | |
Dark Blue |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction