Learn More
OHAUS™ Culture Platform
OHAUS™ Culture Platform designed to hold irregular samples or traditional flask clamps and tube racks.
Brand:  OHAUS™ 30400066
 View more versions of this product
Product Code. 15782479
  • 380.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Culture Platform
18.00 in.
45.7 x 45.7 cm
45.70 cm
45.70 cm
Mat
Stainless Steel
45.70 cm
18.00 in.
18.00 in.
Culture Platform with Mat, 46 X 46 cm