Learn More
Thermo Scientific Chemicals Crystal Violet, pure, high purity biological stain
73.57 € - 273.36 €
Specifications
Chemical Name or Material | Crystal Violet |
---|---|
Melting Point | 215.0°C |
CAS | 548-62-9, 90-94-8 |
Molecular Formula | C25H30ClN3 |
MDL Number | MFCD00011750 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10402041
|
Thermo Scientific Chemicals
229641000 |
100 g | Glass bottle |
73.57 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772461
|
Thermo Scientific Chemicals
229645000 |
500 g | Glass bottle |
273.36 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
Crystal Violet | |
548-62-9, 90-94-8 | |
MFCD00011750 | |
15, 4430 | |
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether | |
Authentic | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
407.99 | |
≥96.0% (TiCl3-titration) (on dried substance) | |
Glass bottle | |
Powder or Crystals |
215.0°C | |
C25H30ClN3 | |
13, 756 | |
Basic Violet 3, C.I. 42555, Gentian Violet | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
9% max. (110°C) | |
Pure | |
Green |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
May cause cancer.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not eat, drink or smoke when using this product.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 208-953-6
missing translation for 'rtecsNumber' : BO9000000
missing translation for 'tsca' : TSCA
- Crystal Violet
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : C.I. Basic Violet 3
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.