missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multiroir™ Transparent Crystal Polystyrene Storage Drawers
View the contents of Multiroir™ Transparent Crystal Polystyrene Storage Drawers clearly due to their construction of crystal clear polystyrene. The drawers also feature a slide assembly, with dividers available to order separately.
Specifications
Color | Transparent |
---|---|
Material | Polystyrene |
Product Type | Storage Drawer |
Quantity | 1 Each |
Description
- Total visibility
- Crystal polystyrene transparent - like glass
- Slide assembly
- Drawer separators available separately
Specifications
Transparent | |
Polystyrene | |
Storage Drawer | |
1 Each |