missing translation for 'onlineSavingsMsg'
Learn More

Multiroir™ Crystal Polystyrene Minimax Storage Boxes

Use Multiroir™ Crystal Polystyrene Minimax Storage Boxes for your storage and transport needs. The clear, glass-like polystyrene enables you to view your contents clearly from the outside. The boxes are delivered complete with a separator kit for additional organization of contents.

Specifications

Color Transparent
Material Polystyrene
Shape Rectangular
Product Type Storage Box
View More Specs

Description

Description

  • Transparent as glass
  • Delivered complete with separator kit
  • Various capacities
Specifications

Specifications

Transparent
Polystyrene
Rectangular
Storage Box
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.