missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Partitions for Cryogenic Vial Boxes
GREENER_CHOICE

Match the partition to your tube requirements and select the appropriate size box. Heathrow Scientific™ Partitions Cryogenic Vial Boxes are constructed from cardboard.

Brand:  Heathrow Scientific™ HS2860H

28.59 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.


Product Code. 11384015

  • 32.30 € / Pack of 12
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with Cryogenic Vial Boxes
  • Cardboard construction
  • Fits 1.5/2.0mL tubes
Specifications

Specifications

White
Cryo Box Partition
12/Pk.
12 mm
Cardboard
81 (1.5 to 2 mL Tubes)
9 x 9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.