missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX Certiprep™ Cryogenic Gloves
SPEX Certiprep™ Cryogenic Gloves are designed to protect hands and arms from ultra-cold materials and liquid nitrogen exposure.
447.00 € - 451.00 €
Specifications
Color | Blue |
---|---|
Material | Fabric |
Powdered | Powder Free |
Product Type | Cryogenic Gloves |
Sterility | Non-sterile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13463530
|
SPEX Certiprep™
6900M |
Medium |
451.00 €
1 pair Qty Price Break Available |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
13424570
|
SPEX Certiprep™
6900L |
Large |
451.00 €
1 pair Qty Price Break Available |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
13473210
|
SPEX Certiprep™
6900S |
Small |
447.00 €
1 pair Qty Price Break Available |
Estimated Shipment: 11-11-2024
Log in to see stock. |
|||||
Description
- Cryogenic gloves specifically designed to protect hands and arms from the hazards associated with liquid nitrogen handling
- Highly recommended for the safe operation of the Freezer/Mill™
- Multi-layer insulation for optimal protection
- Flexible and lightweight to allow excellent dexterity
- Comfortable to wear for extended periods of time
Specifications
Blue | |
Powder Free | |
Non-sterile |
Fabric | |
Cryogenic Gloves |