missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Gloves - Mid-Arm Length

Product Code. 15639316
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY

Specifications

Description Mid-Arm Length
Color Blue
Hand Specific
Size X-Large
For Use With (Application) Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English) 14 in.
Length (Metric) 35.6 cm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'