missing translation for 'onlineSavingsMsg'
Learn More

CREST Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen CREST
Concentration 0.26 mg/mL
Applications Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16850154
View Documents Promotion Details
Proteintech
12439-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16840154
View Documents Promotion Details
Proteintech
12439-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Synovial sarcomas occur most frequently in the extremities around large joints. More than 90% of cases have a recurrent and specific chromosomal translocation, t(X;18)(p11.2;q11.2), in which the 5-prime end of the SS18 gene (MIM 600192) is fused in-frame to the 3-prime end of the SSX1 (MIM 312820), SSX2 (MIM 300192), or SSX4 (MIM 300326) gene. The SS18L1 gene is homologous to SS18.
Specifications

Specifications

CREST
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
O75177, Q8BW22, Q91XJ0
192352, 26039, 269397
CREST Fusion Protein Ag3119
Primary
-20°C
SS18L1
0.26 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CREST, KIAA0693, LP2261, SS18 like protein 1, SS18L1, SYT homolog 1
Ss18l1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications