Learn More
Cole-Parmer™ CPC™ Panel Mount Gasket Couplers
Virtually eliminate any loss of product with this leak-free design
Brand:  Cole-Parmer™ 31603-19
 View more versions of this product
Product Code. 15437333
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
  • For a complete fitting, order one body and one insert.
  • All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifications
Specifications
Panel Mount Gasket
10 Gaskets
NS6 Series Couplers
EPDM