Learn More
COX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12375-1-AP-20UL
 View more versions of this product
Product Code. 16850064
  • 140.00 € / 20µL
Explore more special offers
Description
Description
PTGS1 (COX1, Cyclooxygenase 1) is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes PTGS1, which regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. PTGS1 is thought to be involved in cell-cell signaling and maintaining tissue homeostasis. Alternative splicing of this gene generates two transcript variants. The expression of these two transcripts is differentially regulated by relevant cytokines and growth factors.
Specifications
Specifications
COX2
Polyclonal
Unconjugated
PTGS2
COX 2, COX2, Cyclooxygenase 2, GRIPGHS, hCox 2, PGG/HS, PGH synthase 2, PGHS 2, PHS 2, PHS II, Prostaglandin G/H synthase 2, Prostaglandin H2 synthase 2, PTGS2
Rabbit
Antigen Affinity Chromatography
RUO
19225, 29527, 5743
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35354, P35355, Q05769
PTGS2
COX2 Fusion Protein Ag3025
20 μL
Primary
Mouse, Human, Rat