missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12375-1-AP-150UL
This item is not returnable.
View return policy
Description
PTGS1 (COX1, Cyclooxygenase 1) is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes PTGS1, which regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. PTGS1 is thought to be involved in cell-cell signaling and maintaining tissue homeostasis. Alternative splicing of this gene generates two transcript variants. The expression of these two transcripts is differentially regulated by relevant cytokines and growth factors.Specifications
COX2 | |
Polyclonal | |
Unconjugated | |
PTGS2 | |
COX 2, COX2, Cyclooxygenase 2, GRIPGHS, hCox 2, PGG/HS, PGH synthase 2, PGHS 2, PHS 2, PHS II, Prostaglandin G/H synthase 2, Prostaglandin H2 synthase 2, PTGS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19225, 29527, 5743 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35354, P35355, Q05769 | |
PTGS2 | |
COX2 Fusion Protein Ag3025 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |