Learn More
Kimberly-Clark Professional™ KleenGuard™ A20+ Coveralls
Click to view available options
Size:
Small
3X-Large
Description
- Breathable, silicone-free coveralls, with an elasticated hood, ankle and wrists for comfort, safety and strength
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
- Protects against powders, larger fibres, radioactive dust and chemical splashes
- Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
- Silicone-free, low lint and treated to be antistatic - to EN 1149-5
- Single-use protective clothing to improve safety and comfort in high-risk areas. Reduces contamination in critical areas
- Offers complete freedom of movement with comfortable body design
- Ideal for working in general manufacturing and maintenance
- Tripled-stitched seams help to protect against tearing with an easy-fastening snap front
Specifications
Specifications
Product Type | Single-use Coveralls |
Size | 3X-Large |
Gender | Unisex |
Disposable | Single-use |
Sterility | Non-sterile |
Product Content Correction
Product Title
Kimberly-Clark Professional™ KleenGuard™ A20+ Coveralls >
Spot an opportunity for improvement?Share a Content Correction