Learn More
Masterflex™ Corrugated FEP Tubing
High flexibility and chemical resistance.
Brand: Masterflex™ 06407-60
This item is not returnable.
View return policy
Description
- Corrugations prevent kinking and help tubing retain shape without limiting pressure or vacuum abilities.
- Custom extruded.
- Can be cut to desired length.
- Sold in 5-ft lengths.
- Alternating sections of corrugated (10-in. length), and noncorrugated (2-in. length), with noncorrugated 1-in. lengths at both ends.
- Accepts compression fittings.
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;NSF-61;USP;3A;European Pharmacopoeia;NSF-51 | |
FEP | |
1.5 m | |
-184°C | |
250 psi | |
204 °C |
5.6 mm | |
6.4 mm | |
0.41275 mm | |
Translucent | |
Withstands 27-in. Hg vacuum |