missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PTFE-Coated Magnetic Stir Bars
For use in Corning Hotplate Stirrers
33.49 € - 50.71 €
Product Code | Brand | Diameter (English) | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity & Availability | ||
10183943
|
Corning™
400430 |
0.37 in. | 9.5 mm | 2 in. | 50.8 mm |
39.36 €
Each |
In Stock
Log in to see stock. |
||
10628653
|
Corning™
401435 |
0.37 in. | 9.5 mm | 1 in. | 25.4 mm |
33.49 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||
10626953
|
Corning™
409000 |
0.75 in. | 19 mm | 3 in. | 76.2 mm |
50.71 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||
Description
Corning magnetic stir bars are polytetrafluorethylene (PTFE)-coated for increased chemical resistance.Specifications
Magnet | |
1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title