missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Insectagro™ Sf9 Serum-free/Protein-free Medium, 1X, With L-glutamine
Formulated to support propagation of Sf9 insect cells in culture.
Brand: Corning™ 13-410-CV
This item is not returnable.
View return policy
Description
- Satisfies demands of Sf9 and Sf21 cultures
- Cultured in non-humidified, non-CO2 incubators at 27°C
- Display both monolayer and suspension culture qualities
- With their fast doubling times of 18 to 22 hrs, Sf9 cells are easily scaled up to large cultures using bioreactors
- Capable of expressing full-length proteins
- Have been reported to produce certain proteins up to 20% of total protein
Order Info
Liquid and powdered media are packaged in PET with tamper-evident seals; shipped at ambient temperature. Certificates of Analysis for each lot are available from Mediatech upon request.
Compliance
Liquid products are tested for sterility according to FDA Guidelines for the Manufacture of In Vitro Diagnostic Products (January 1994) for sterile labeling.
Specifications
2° to 8°C | |
Liquid | |
Sterile | |
0.1364 EU/ mL | |
6.7 | |
RUO |
1 X | |
Insect Cell Serum Free Medium (SFM), Protein-free | |
Glutamine | |
Insect cell culture | |
6 x 500 mL | |
Originated at USDA Insect Pathology Laboratory from IPLBSF-21 cell line, derived from pupal ovarian tissue of fall army worm |
For Research Use Only.