Learn More
Corning™ Human Umbilical Vein Endothelial Cells (HUVEC-2)
Used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents
Brand:  Corning™ 354151
Product Code. 10683493
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Corning™ HUVEC-2 cells are qualified for use in Corning™ BioCoat™ Angiogenesis Systems: Endothelial Cell Migration. The cells may be used in Corning BioCoat Endothelial Cell Invasion and Tube Formation assays. Packaged in individual cryogenic vials containing >5x105 cells.
  • Excellent viability
  • Each vial of Corning™ HUVEC-2 cells contain 5x105 cells that have been cryopreserved at passage number 2 in medium containing 10% DMSO.
  • Lot-to-lot consistency
  • Tested for the ability to produce a robust migratory response in the presence of VEGF and FBS.
  • Prescreened for Angiogenesis studies
  • Qualified for use in Corning™ BioCoat Angiogenesis Systems: Endothelial Cell Migration. May also be used in Corning™ BioCoat Endothelial Cell Invasion and Tube Formation assays.
  • Positive immunohistochemical staining for von Willebrand factor (vWf) and CD31 antigen
  • Positive Dil-Ac-LDL uptake
  • Negative mycoplasma, HIV-1, hepatitis B and C, bacteria, yeast, and fungi
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Corning™ BioCoat™ Angiogenesis System
Human Umbilical Vein Endothelial Cells
Human Umbilical Vein
Used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents
>5 x 105 Cells