missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) without Glutamine
Guaranteed low endotoxin levels ≤0.025EU/mL.
168.53 €
Specifications
Content And Storage | 2° to 8°C |
---|---|
Form | Liquid |
Sterility | Sterile |
pH | 7 to 7.4 |
Product Code | Brand | Without Additives | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Without Additives | Quantity | Price | Quantity & Availability | ||||
15303571
|
Corning™
17-305-CV |
No Glutamine, No Phenol Red | 6 x 500 mL |
168.53 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. This research indicated that additions made to BME aided cell propagation. MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayer, such as fibroblast. This formulation contains sodium bicarbonate and does not contain L-glutamine.Specifications
2° to 8°C | |
Liquid | |
Sterile | |
7 to 7.4 |